2-(Diethylamino)-4-pyrimidinecarbonitrile 95%

Lieferant: Apollo Scientific

OR33537-1G
APOSOR33537-1GEA 874 EUR
APOSOR33537-1G
2-(Diethylamino)-4-pyrimidinecarbonitrile 95%
2-(Diethylamino)-4-pyrimidinecarbonitrile
Formel: C₉H₁₂N₄
Molecular Weight: 176,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15202259
CAS-Nummer: 75825-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR