2-Iodnicotinonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR3354-250MG
APOSOR3354-250MGEA
21.9
EUR
APOSOR3354-250MG
2-Iodnicotinonitril 98%
2-Iodnicotinonitril
Formel:
C₆H₃IN₂ Schmelzpunkt: 187…189 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436053 CAS-Nummer: 490039-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...