8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride 95%

Lieferant: Apollo Scientific

OR33540-1G
APOSOR33540-1GEA 270 EUR
APOSOR33540-1G
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride 95%
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride
Formel: C₇H₁₄ClNOS
Molecular Weight: 195.71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140314
CAS-Nummer: 1221791-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR