4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone 95%
Lieferant: Apollo Scientific
OR33541-1G
APOSOR33541-1GEA
427
EUR
APOSOR33541-1G
4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone 95%
4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone
Formel:
C₁₀H₁₃NO Molecular Weight: 163,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037462 CAS-Nummer: 16236-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...