1-(4-Amino-3-methoxyphenyl)-1-ethanol 95%

Lieferant: Apollo Scientific

OR33545-1G
APOSOR33545-1GEA 671 EUR
APOSOR33545-1G
1-(4-Amino-3-methoxyphenyl)-1-ethanol 95%
1-(4-Amino-3-methoxyphenyl)-1-ethanol
Formel: C₉H₁₃NO₂
Molecular Weight: 167.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140262
CAS-Nummer: 1221792-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR