1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride 95%

Lieferant: Apollo Scientific

OR33546-1G
APOSOR33546-1GEA 336 EUR
APOSOR33546-1G
1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride 95%
1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride
Formel: C₈H₁₆ClNOS
Molecular Weight: 209.74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140323
CAS-Nummer: 1221792-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR