6,7-Dihydro-1H-cyclopenta[d]pyrimidine-2,4(3H,5H)-dione 95%

Lieferant: Apollo Scientific

OR33547-1G
APOSOR33547-1GEA 197 EUR
APOSOR33547-1G
6,7-Dihydro-1H-cyclopenta[d]pyrimidine-2,4(3H,5H)-dione 95%
6,7-Dihydro-1H-cyclopenta[d]pyrimidine-2,4(3H,5H)-dione
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466420
CAS-Nummer: 5466-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR