1-Methyl-1H-1,2,3-benzotriazol-5-amine 97%

Lieferant: Apollo Scientific

OR33555-1G
APOSOR33555-1GEA 1070 EUR
APOSOR33555-1G
1-Methyl-1H-1,2,3-benzotriazol-5-amine 97%
1-Methyl-1H-1,2,3-benzotriazol-5-amine
Formel: C₇H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18651767
CAS-Nummer: 27799-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR