N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide 95%

Lieferant: Apollo Scientific

OR33557-1G
APOSOR33557-1GEA 1120 EUR
APOSOR33557-1G
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide 95%
N-(4,5-Dimethyl-1H-imidazol-2-yl)acetamide
Formel: C₇H₁₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09909729
CAS-Nummer: 40639-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR