N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide 95%

Lieferant: Apollo Scientific

OR33558-1G
APOSOR33558-1GEA 270 EUR
APOSOR33558-1G
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide 95%
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide
Formel: C₆H₁₁BrN₂S
Molecular Weight: 223.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651784
CAS-Nummer: 849004-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR