5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one, tech

Lieferant: Apollo Scientific

OR33566-1G
APOSOR33566-1GEA 1570 EUR
APOSOR33566-1G
5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one, tech
5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one
Formel: C₉H₉N₃O
Molecular Weight: 175,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501765
CAS-Nummer: 1092344-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR