tert-Butyl-1,2-diazepane-1-carboxylate 95%

Lieferant: Apollo Scientific

OR33567-1G
APOSOR33567-1GEA 874 EUR
APOSOR33567-1G
tert-Butyl-1,2-diazepane-1-carboxylate 95%
tert-Butyl-1,2-diazepane-1-carboxylate
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09607901
CAS-Nummer: 443295-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR