5-Chloro-2-oxo-1,2-dihydro-3-pyridinecarbaldehyde 95%

Lieferant: Apollo Scientific

OR33568-1G
APOSOR33568-1GEA 1300 EUR
APOSOR33568-1G
5-Chloro-2-oxo-1,2-dihydro-3-pyridinecarbaldehyde 95%
5-Chloro-2-oxo-1,2-dihydro-3-pyridinecarbaldehyde
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972232
CAS-Nummer: 614732-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR