4-Amino-3-(4-bromphenyl)-1H-pyrazol 95%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Bromophenyl)-1H-pyrazol-4-amine
OR33574-1G
APOSOR33574-1GEA
538
EUR
APOSOR33574-1G
4-Amino-3-(4-bromphenyl)-1H-pyrazol 95%
4-Amino-3-(4-bromophenyl)-1H-pyrazole
Formel:
C₉H₈BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758068 CAS-Nummer: 91857-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...