3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific

OR33576-1G
APOSOR33576-1GEA 37.7 EUR
APOSOR33576-1G
3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 95%
3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₄BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11100204
CAS-Nummer: 866546-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR