Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2,5,6-trichloropyrimidine-4-carboxylate

OR33577-1G
APOSOR33577-1GEA 122 EUR
APOSOR33577-1G
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate 97%
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate
Formel: C₆H₃Cl₃N₂O₂
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025876
CAS-Nummer: 89284-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR