N-(2-Formyl-3-methoxyphenyl)-2,2-dimethylpropanamide 95%
Lieferant: Apollo Scientific
OR33580-1G
APOSOR33580-1GEA
671
EUR
APOSOR33580-1G
N-(2-Formyl-3-methoxyphenyl)-2,2-dimethylpropanamide 95%
N-(2-Formyl-3-methoxyphenyl)-2,2-dimethylpropanamide
Formel:
C₁₃H₁₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09955251 CAS-Nummer: 82673-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...