1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone 95%
Lieferant: Apollo Scientific
OR33583-1G
APOSOR33583-1GEA
671
EUR
APOSOR33583-1G
1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone 95%
1-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1-ethanone
Formel:
C₁₅H₁₂N₂O Molecular Weight: 236,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09759184 CAS-Nummer: 122643-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...