tert-Butyl 9-cyano-3-azaspiro[5.5]undecane-3-carboxylate, tech
Lieferant: Apollo Scientific
OR33588-1G
APOSOR33588-1GEA
1570
EUR
APOSOR33588-1G
tert-Butyl 9-cyano-3-azaspiro[5.5]undecane-3-carboxylate, tech
tert-Butyl 9-cyano-3-azaspiro[5.5]undecane-3-carboxylate
Formel:
C₁₆H₂₆N₂O₂ Molecular Weight: 278,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18426549 CAS-Nummer: 1281872-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...