4-(1,3-Benzoxazol-5-yl)benzaldehyde 95%

Lieferant: Apollo Scientific

OR33591-1G
APOSOR33591-1GEA 538 EUR
APOSOR33591-1G
4-(1,3-Benzoxazol-5-yl)benzaldehyde 95%
4-(1,3-Benzoxazol-5-yl)benzaldehyde
Formel: C₁₄H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18642384
CAS-Nummer: 1008361-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR