2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile 97%
Lieferant: Apollo Scientific
OR33597-1G
APOSOR33597-1GEA
180
EUR
APOSOR33597-1G
2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile 97%
2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile
Formel:
C₁₁H₁₃ClN₂ Molecular Weight: 208.69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10014479 CAS-Nummer: 1018265-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...