4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride 97%

Lieferant: Apollo Scientific

OR33599-1G
APOSOR33599-1GEA 1120 EUR
APOSOR33599-1G
4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride 97%
4-{1H-Imidazo[4,5-b]pyridin-1-yl}piperidine hydrochloride
Formel: C₁₁H₁₅ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22126107
CAS-Nummer: 1370587-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR