4-Chlorpicolinsäurehydrazid 98%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-(hydrazinecarbonyl)pyridine

OR3360-1G
APOSOR3360-1GEA 65.4 EUR
APOSOR3360-1G
4-Chlorpicolinsäurehydrazid 98%
4-Chlorpicolinsäurehydrazid
Formel: C₆H₆ClN₃O
Molecular Weight: 171,5843 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01704394
CAS-Nummer: 73771-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR