Methyl-3-methyl-1-benzofuran-2-carboxylate 97%
Lieferant: Apollo Scientific
OR33601-1G
APOSOR33601-1GEA
435
EUR
APOSOR33601-1G
Methyl-3-methyl-1-benzofuran-2-carboxylate 97%
Methyl-3-methyl-1-benzofuran-2-carboxylate
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09270117 CAS-Nummer: 2076-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...