(5-Methylpyridin-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR33603-1G
APOSOR33603-1GEA 197 EUR
APOSOR33603-1G
(5-Methylpyridin-2-yl)methanol 95%
(5-Methylpyridin-2-yl)methanol
Formel: C₇H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09999992
CAS-Nummer: 22940-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR