1-Benzyl-4-(chloromethyl)-1H-1,2,3-triazole 97%

Lieferant: Apollo Scientific

OR33608-1G
APOSOR33608-1GEA 627 EUR
APOSOR33608-1G
1-Benzyl-4-(chloromethyl)-1H-1,2,3-triazole 97%
1-Benzyl-4-(chloromethyl)-1H-1,2,3-triazole
Formel: C₁₀H₁₀ClN₃
Molecular Weight: 207.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16748189
CAS-Nummer: 70380-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR