4-Chloro-2-methyl-5-[(6-methylpyridin-3-yl)oxy]-2,3-dihydropyridazin-3-one 97%
Lieferant: Apollo Scientific
OR33610-1G
APOSOR33610-1GEA
538
EUR
APOSOR33610-1G
4-Chloro-2-methyl-5-[(6-methylpyridin-3-yl)oxy]-2,3-dihydropyridazin-3-one 97%
4-Chloro-2-methyl-5-[(6-methylpyridin-3-yl)oxy]-2,3-dihydropyridazin-3-one
Formel:
C₁₁H₁₀ClN₃O₂ Molecular Weight: 251,669 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28016138 CAS-Nummer: 1597781-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...