Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate 97%

Lieferant: Apollo Scientific

OR33611-1G
APOSOR33611-1GEA 336 EUR
APOSOR33611-1G
Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate 97%
Methyl (2Z)-3-(2,6-dichlorophenyl)-2-acetamidoprop-2-enoate
Formel: C₁₂H₁₁Cl₂NO₃
Molecular Weight: 288.127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28016143
CAS-Nummer: 1598370-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR