tert-Butyl-4-(3-cyanophenyl)piperazine-1-carboxylate 95%
Lieferant: Apollo Scientific
OR33613-1G
APOSOR33613-1GEA
427
EUR
APOSOR33613-1G
tert-Butyl-4-(3-cyanophenyl)piperazine-1-carboxylate 95%
tert-Butyl-4-(3-cyanophenyl)piperazine-1-carboxylate
Formel:
C₁₆H₂₁N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042238 CAS-Nummer: 807624-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...