3-Amino-6-bromo-8-methyl-3,4-dihydroquinazolin-4-one, tech
Lieferant: Apollo Scientific
OR33614-1G
APOSOR33614-1GEA
1570
EUR
APOSOR33614-1G
3-Amino-6-bromo-8-methyl-3,4-dihydroquinazolin-4-one, tech
3-Amino-6-bromo-8-methyl-3,4-dihydroquinazolin-4-one
Formel:
C₉H₈BrN₃O Molecular Weight: 254,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11501800 CAS-Nummer: 1092345-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...