2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol 95%

Lieferant: Apollo Scientific

Synonyme: 2-[1-Benzyl-4-(hydroxymethyl)pyrrolidin-3-yl]phenol

OR33618-1G
APOSOR33618-1GEA 894 EUR
APOSOR33618-1G
2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol 95%
2-[1-Benzyl-4-(hydroxymethyl)tetrahydro-1H-pyrrol-3-yl]benzenol
Formel: C₁₈H₂₁NO₂
Molecular Weight: 283,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10758080
CAS-Nummer: 1033463-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR