4-Amino-3-(3-bromphenyl)-1H-pyrazol 95%
Lieferant: Apollo Scientific
Synonyme:
3-(3-Bromophenyl)-1H-pyrazol-4-amine
OR33619-1G
APOSOR33619-1GEA
538
EUR
APOSOR33619-1G
4-Amino-3-(3-bromphenyl)-1H-pyrazol 95%
4-Amino-3-(3-bromophenyl)-1H-pyrazole
Formel:
C₉H₈BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100205 CAS-Nummer: 1171492-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...