Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride 95%

Lieferant: Apollo Scientific

OR33623-1G
APOSOR33623-1GEA 488 EUR
APOSOR33623-1G
Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride 95%
Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride
Formel: C₈H₁₆ClNO₂
Molecular Weight: 193,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10758087
CAS-Nummer: 1171925-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR