3-Iodimidazo[1,2-a]pyridin-2-carbonsäure 95%
Lieferant: Apollo Scientific
OR33624-1G
APOSOR33624-1GEA
538
EUR
APOSOR33624-1G
3-Iodimidazo[1,2-a]pyridin-2-carbonsäure 95%
3-Iodimidazo[1,2-a]pyridin-2-carbonsäure
Formel:
C₈H₅IN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758089 CAS-Nummer: 1033463-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...