3-Amino-1,2,3,4-tetrahydroquinolin-2-one hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR33629-1G
APOSOR33629-1GEA 488 EUR
APOSOR33629-1G
3-Amino-1,2,3,4-tetrahydroquinolin-2-one hydrochloride 95%
3-Amino-1,2,3,4-tetrahydroquinolin-2-one hydrochloride
Formel: C₉H₁₀N₂O·ClH
Molecular Weight: 198,649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09999258
CAS-Nummer: 35849-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR