6-Chloro-3,4-dihydro-2H-1,4-benzoxazine 95%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine
OR33630-1G
APOSOR33630-1GEA
671
EUR
APOSOR33630-1G
6-Chloro-3,4-dihydro-2H-1,4-benzoxazine 95%
6-Chloro-3,4-dihydro-2H-1,4-benzoxazine
Formel:
C₈H₈ClNO Molecular Weight: 169,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544339 CAS-Nummer: 70558-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...