3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol 95%

Lieferant: Apollo Scientific

OR33631-1G
APOSOR33631-1GEA 1330 EUR
APOSOR33631-1G
3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol 95%
3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848182
CAS-Nummer: 915160-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR