6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one 95%

Lieferant: Apollo Scientific

OR33632-1G
APOSOR33632-1GEA 894 EUR
APOSOR33632-1G
6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one 95%
6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one
Formel: C₇H₅BrN₂O₂
Molecular Weight: 229,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025916
CAS-Nummer: 335032-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR