(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR33634-1G
APOSOR33634-1GEA 538 EUR
APOSOR33634-1G
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol 95%
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol
Formel: C₅H₆Br₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12025921
CAS-Nummer: 881997-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR