Ethyl 1H-indole-6-carboxylate 97%
Lieferant: Apollo Scientific
OR33643-1G
APOSOR33643-1GEA
0
EUR
APOSOR33643-1G
Ethyl 1H-indole-6-carboxylate 97%
Ethyl 1H-indole-6-carboxylate
Formel:
C₁₁H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06205072 CAS-Nummer: 50820-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...