5-Chloro-2-methylnicotinic acid 97%
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-2-methyl-3-pyridinecarboxylic acid, 5-Chloro-2-methylpyridine-3-carboxylic acid
OR33645-1G
APOSOR33645-1GEA
59.6
EUR
APOSOR33645-1G
5-Chloro-2-methylnicotinic acid 97%
5-Chloro-2-methylnicotinic acid
Formel:
C₇H₆ClNO₂ Molecular Weight: 171,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11557228 CAS-Nummer: 1092286-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...