Methyl 5-methoxy-3-methyl-1H-indole-2-carboxylate, tech
Lieferant: Apollo Scientific
OR33647-1G
APOSOR33647-1GEA
1570
EUR
APOSOR33647-1G
Methyl 5-methoxy-3-methyl-1H-indole-2-carboxylate, tech
Methyl 5-methoxy-3-methyl-1H-indole-2-carboxylate
Formel:
C₁₂H₁₃NO₃ Molecular Weight: 219,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06496116 CAS-Nummer: 480996-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...