[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR33648-250MG
APOSOR33648-250MGEA 179 EUR
APOSOR33648-250MG
[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride
[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride
Formel: C₈H₈N₂O₂·ClH
Molecular Weight: 200.622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660999
CAS-Nummer: 1255717-94-2
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR