Ethylimidazo[1,2-a]pyridin-7-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR3366-1G
APOSOR3366-1GEA
30.6
EUR
APOSOR3366-1G
Ethylimidazo[1,2-a]pyridin-7-carboxylat
Ethylimidazo[1,2-a]pyridin-7-carboxylat
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190,1986 g/mol Schmelzpunkt: 87…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436046 CAS-Nummer: 372147-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...