Methyl 5-(aminomethyl)-2-cyclopropyl-pyrazole-3-carboxylate hydrochloride

Lieferant: Apollo Scientific
Achtung

OR33661-100MG OR33661-1G OR33661-250MG
APOSOR33661-100MGEA 48 EUR
APOSOR33661-100MG APOSOR33661-1G APOSOR33661-250MG
Methyl 5-(aminomethyl)-2-cyclopropyl-pyrazole-3-carboxylate hydrochloride
Methyl 5-(aminomethyl)-2-cyclopropyl-pyrazole-3-carboxylate hydrochloride
Formel: C₉H₁₃N₃O₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD31812084
CAS-Nummer: 2391987-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR