4-Bromo-N,N-dimethylbenzylamine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromophenyl)-N,N-dimethylmethanamine, 1-(4-Bromophenyl)-N,N-dimethylmethylamine

OR3370-10G OR3370-1G OR3370-5G
APOSOR3370-10GEA 270 EUR
APOSOR3370-10G APOSOR3370-1G APOSOR3370-5G
4-Bromo-N,N-dimethylbenzylamine 98%
4-Bromo-N,N-dimethylbenzylamine
Formel: C₉H₁₂BrN
Molecular Weight: 214.102 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00032473
CAS-Nummer: 6274-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR