2-(Piperazin-1-yl)-N-(pyridin-3-yl)acetamide trihydrochloride

Lieferant: Apollo Scientific
Achtung

OR3376-1G
APOSOR3376-1GEA 0 EUR
APOSOR3376-1G
2-(Piperazin-1-yl)-N-(pyridin-3-yl)acetamide trihydrochloride
2-(Piperazin-1-yl)-N-(pyridin-3-yl)acetamide trihydrochloride
Formel: C₁₁H₁₆N₄O·3ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03844727
CAS-Nummer: 808764-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR