1-Pyridin-3-ylmethylpiperazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Pyridin-3-ylmethyl)piperazine
OR3380-1G
APOSOR3380-1GEA
27.7
EUR
APOSOR3380-1G
1-Pyridin-3-ylmethylpiperazine 97%
1-Pyridin-3-ylmethylpiperazine
Formel:
C₁₀H₁₅N₃ Molecular Weight: 177,25 g/mol Siedepunkt: 84…86 °C/0,1mmHg Dichte: 1,1 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862552 CAS-Nummer: 39244-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...