1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3,4,6-Tetra-O-acetyl-2-amino-2-deoxy-beta-D-glucopyranose hydrochloride, (2S,3R,4R,5S,6R)-6-(Acetoxymethyl)-3-aminotetrahydro-2H-pyran-2,4,5-triyl triacetate hydrochloride

OR3381-250MG
APOSOR3381-250MGEA 21.9 EUR
APOSOR3381-250MG
1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride 98%
1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
Formel: C₁₄H₂₂ClNO₉
Molecular Weight: 383,778 g/mol
Schmelzpunkt: >225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075204
CAS-Nummer: 10034-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR