5-Chloro-2-methoxyphenylhydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-hydrazino-1-methoxybenzene hydrochloride, 4-Chloro-2-hydrazinoanisole hydrochloride

OR3385-1G
APOSOR3385-1GEA 125 EUR
APOSOR3385-1G
5-Chloro-2-methoxyphenylhydrazine hydrochloride
5-Chloro-2-methoxyphenylhydrazine hydrochloride
Formel: C₇H₉ClN₂O·ClH
Molecular Weight: 209,073 g/mol
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03196500
CAS-Nummer: 5446-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR